Learn More
Gilson Company Inc SIEVE 3 SS FH NO12
Catalog No. 50871298
Encompass_Preferred
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Full height; 3 in. dia.; No. 12; Sieve opening; 1.7mm
Catalog No. 50-871-298 Supplier Gilson Company Inc Supplier No. V3SF #12
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Full height; 3 in. dia.; No. 12; Sieve opening; 1.7mm