Learn More
Gilson Company Inc SIEVE 3 SS FH NO14
Catalog No. 50871299
Encompass_Preferred
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Full height; 3 in. dia.; No. 14; Sieve opening; 1.4mm
Catalog No. 50-871-299 Supplier Gilson Company Inc Supplier No. V3SF #14
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Full height; 3 in. dia.; No. 14; Sieve opening; 1.4mm