Learn More
Gilson Company Inc SIEVE 3 SS FH NO16
Catalog No. 50871300
Encompass_Preferred
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Full height; 3 in. dia.; No. 16; Sieve opening; 1.18mm
Catalog No. 50-871-300 Supplier Gilson Company Inc Supplier No. V3SF #16
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Full height; 3 in. dia.; No. 16; Sieve opening; 1.18mm