Learn More
Gilson Company Inc SIEVE 3 SS FH NO30
Catalog No. 50871304
Encompass_Preferred
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Full height; 3 in. dia.; No. 30; Sieve opening; 600um
Catalog No. 50-871-304 Supplier Gilson Company Inc Supplier No. V3SF #30
Add to Cart
Add to Cart
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Full height; 3 in. dia.; No. 30; Sieve opening; 600um