Learn More
Gilson Company Inc SIEVE 3 SS FH NO4
Catalog No. 50871292
Encompass_Preferred
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Full height; 3 in. dia.; No. 4; Sieve opening; 4.75mm
Catalog No. 50-871-292 Supplier Gilson Company Inc Supplier No. V3SF #4
Add to Cart
Add to Cart
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Full height; 3 in. dia.; No. 4; Sieve opening; 4.75mm