Learn More
Gilson Company Inc SIEVE 3 SS FH NO450
Catalog No. 50871321
Encompass_Preferred
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Full height; 3 in. dia.; No. 450; Sieve opening; 32um
Catalog No. 50-871-321 Supplier Gilson Company Inc Supplier No. V3SF #450
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Full height; 3 in. dia.; No. 450; Sieve opening; 32um