Learn More
Gilson Company Inc SIEVE 3 SS FH NO50
Catalog No. 50871308
Encompass_Preferred
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Full height; 3 in. dia.; No. 45; Sieve opening; 300um
Catalog No. 50-871-308 Supplier Gilson Company Inc Supplier No. V3SF #50
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Full height; 3 in. dia.; No. 45; Sieve opening; 300um