Learn More
Gilson Company Inc SIEVE 3 SS FH NO6
Supplier:  Gilson Company Inc V3SF #6
Encompass_Preferred
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Full height; 3 in. dia.; No. 7; Sieve opening; 2.8mm
Catalog No. 50-871-294