Learn More
Gilson Company Inc SIEVE 3 SS FH NO7
Catalog No. 50871295
Encompass_Preferred
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Full height; 3 in. dia.; No. 6; Sieve opening; 3.35mm
Catalog No. 50-871-295 Supplier Gilson Company Inc Supplier No. V3SF #7
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Full height; 3 in. dia.; No. 6; Sieve opening; 3.35mm