Learn More
Gilson Company Inc 3in (76mm) diameter Stainless Steel full height Pan
Catalog No. 50871327
Encompass_Preferred
3in (76mm) diameter Stainless Steel full height Pan for ASTM E11 3in Test Sieves is placed at the bottom of a sieve stack to collect fines passing through the bottom sieve. Features: Stainless Steel Pan for fines collection, For use with 3in (76mm) diameter ASTM test sieves. Included Items: 3in All Stainless Sieve Pan, Full Height.
Catalog No. 50-871-327 Supplier Gilson Company Inc Supplier No. V3SFXPN
3in (76mm) diameter Stainless Steel full height Pan for ASTM E11 3in Test Sieves is placed at the bottom of a sieve stack to collect fines passing through the bottom sieve. Features: Stainless Steel Pan for fines collection, For use with 3in (76mm) diameter ASTM test sieves. Included Items: 3in All Stainless Sieve Pan, Full Height.