Learn More
Gilson Company Inc 3in (76mm) diameter ASTM E11 Test Sieve, No. 100
Catalog No. 50871349
Encompass_Preferred
3in (76mm) diameter ASTM E11 Test Sieve has stainless steel woven-wire cloth with No. 100 (150µm) openings and half height stainless steel frame. Overall height is 1-1/4in and stacked height is 5/8in. Features: Individually serial numbered, Compliant with ASTM E11 requirements for Compliance Sieves, Included Items: 3” Sieve, All Stainless, Half Height, No. 100, Certificate of Conformance to ASTM E11 requirements.
Catalog No. 50-871-349 Supplier Gilson Company Inc Supplier No. V3SH #100
3in (76mm) diameter ASTM E11 Test Sieve has stainless steel woven-wire cloth with No. 100 (150µm) openings and half height stainless steel frame. Overall height is 1-1/4in and stacked height is 5/8in. Features: Individually serial numbered, Compliant with ASTM E11 requirements for Compliance Sieves, Included Items: 3” Sieve, All Stainless, Half Height, No. 100, Certificate of Conformance to ASTM E11 requirements.