Learn More
Gilson Company Inc SIEVE 3 SS HH NO120
Catalog No. 50871350
Encompass_Preferred
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Half height; 3 in. dia.; No. 120; Sieve opening; 125um
Catalog No. 50-871-350 Supplier Gilson Company Inc Supplier No. V3SH #120
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Half height; 3 in. dia.; No. 120; Sieve opening; 125um