Learn More
Gilson Company Inc SIEVE 3 SS HH NO140
Catalog No. 50871351
Encompass_Preferred
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Half height; 3 in. dia.; No. 140; Sieve opening; 106um
Catalog No. 50-871-351 Supplier Gilson Company Inc Supplier No. V3SH #140
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Half height; 3 in. dia.; No. 140; Sieve opening; 106um