Learn More
Gilson Company Inc SIEVE 3 SS HH NO170
Catalog No. 50871352
Encompass_Preferred
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Half height; 3 in. dia.; No. 170; Sieve opening; 90um
Catalog No. 50-871-352 Supplier Gilson Company Inc Supplier No. V3SH #170
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Half height; 3 in. dia.; No. 170; Sieve opening; 90um