Learn More
Gilson Company Inc SIEVE 3 SS HH NO18
Catalog No. 50871338
Encompass_Preferred
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Half height; 3 in. dia.; No. 18; Sieve opening; 850um
Catalog No. 50-871-338 Supplier Gilson Company Inc Supplier No. V3SH #18
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Half height; 3 in. dia.; No. 18; Sieve opening; 850um