Learn More
Gilson Company Inc SIEVE 3 SS HH NO20
Catalog No. 50871339
Encompass_Preferred
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Half height; 3 in. dia.; No. 18; Sieve opening; 1mm
Catalog No. 50-871-339 Supplier Gilson Company Inc Supplier No. V3SH #20
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Half height; 3 in. dia.; No. 18; Sieve opening; 1mm