Learn More
Gilson Company Inc SIEVE 3 SS HH NO25
Catalog No. 50871340
Encompass_Preferred
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Half height; 3 in. dia.; No. 25; Sieve opening; 710um
Catalog No. 50-871-340 Supplier Gilson Company Inc Supplier No. V3SH #25
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Half height; 3 in. dia.; No. 25; Sieve opening; 710um