Learn More
Gilson Company Inc SIEVE 3 SS HH NO270
Catalog No. 50871355
Encompass_Preferred
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Half height; 3 in. dia.; No. 270; Sieve opening; 53um
Catalog No. 50-871-355 Supplier Gilson Company Inc Supplier No. V3SH #270
Add to Cart
Add to Cart
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Half height; 3 in. dia.; No. 270; Sieve opening; 53um