Learn More
Gilson Company Inc SIEVE 3 SS HH NO30
Catalog No. 50871341
Encompass_Preferred
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Half height; 3 in. dia.; No. 30; Sieve opening; 600um
Catalog No. 50-871-341 Supplier Gilson Company Inc Supplier No. V3SH #30
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Half height; 3 in. dia.; No. 30; Sieve opening; 600um