Learn More
Gilson Company Inc SIEVE 3 SS HH NO4
Catalog No. 50871329
Encompass_Preferred
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Half height; 3 in. dia.; No. 4; Sieve opening; 4.75mm
Catalog No. 50-871-329 Supplier Gilson Company Inc Supplier No. V3SH #4
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Half height; 3 in. dia.; No. 4; Sieve opening; 4.75mm