Learn More
Gilson Company Inc SIEVE 3 SS HH NO400
Catalog No. 50871357
Encompass_Preferred
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Half height; 3 in. dia.; No. 400; Sieve opening; 38um
Catalog No. 50-871-357 Supplier Gilson Company Inc Supplier No. V3SH #400
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Half height; 3 in. dia.; No. 400; Sieve opening; 38um