Learn More
Gilson Company Inc SIEVE 3 SS HH NO5
Catalog No. 50871330
Encompass_Preferred
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Half height; 3 in. dia.; No. 5; Sieve opening; 4mm
Catalog No. 50-871-330 Supplier Gilson Company Inc Supplier No. V3SH #5
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Half height; 3 in. dia.; No. 5; Sieve opening; 4mm