Learn More
Gilson Company Inc SIEVE 3 SS HH NO50
Catalog No. 50871345
Encompass_Preferred
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Half height; 3 in. dia.; No. 50; Sieve opening; 300um
Catalog No. 50-871-345 Supplier Gilson Company Inc Supplier No. V3SH #50
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Half height; 3 in. dia.; No. 50; Sieve opening; 300um