Learn More
Gilson Company Inc SIEVE 3 SS HH NO60
Catalog No. 50871346
Encompass_Preferred
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Half height; 3 in. dia.; No. 60; Sieve opening; 250um
Catalog No. 50-871-346 Supplier Gilson Company Inc Supplier No. V3SH #60
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Half height; 3 in. dia.; No. 60; Sieve opening; 250um