Learn More
Gilson Company Inc SIEVE 3 SS HH NO635
Supplier:  Gilson Company Inc V3SH #635
Encompass_Preferred
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Half height; 3 in. dia.; No. 635; Sieve opening; 20um
Catalog No. 50-871-360