Learn More
Gilson Company Inc SIEVE 3 SS HH NO7
Catalog No. 50871332
Encompass_Preferred
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Half height; 3 in. dia.; No. 7; Sieve opening; 2.8mm
Catalog No. 50-871-332 Supplier Gilson Company Inc Supplier No. V3SH #7
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Half height; 3 in. dia.; No. 7; Sieve opening; 2.8mm