Learn More
Gilson Company Inc SIEVE 3 SS HH NO70
Catalog No. 50871347
Encompass_Preferred
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Half height; 3 in. dia.; No. 70; Sieve opening; 212um
Catalog No. 50-871-347 Supplier Gilson Company Inc Supplier No. V3SH #70
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Half height; 3 in. dia.; No. 70; Sieve opening; 212um