Learn More
Gilson Company Inc SIEVE 3 SS HH NO8
Catalog No. 50871333
Encompass_Preferred
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Half height; 3 in. dia.; No. 8; Sieve opening; 2.36mm
Catalog No. 50-871-333 Supplier Gilson Company Inc Supplier No. V3SH #8
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Half height; 3 in. dia.; No. 8; Sieve opening; 2.36mm