Learn More
Gilson Company Inc SIEVE 3 SS HH NO80
Catalog No. 50871348
Encompass_Preferred
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Half height; 3 in. dia.; No. 80; Sieve opening; 180um
Catalog No. 50-871-348 Supplier Gilson Company Inc Supplier No. V3SH #80
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Half height; 3 in. dia.; No. 80; Sieve opening; 180um