Learn More
Gilson Company Inc SIEVE ACRYLIC 3 SS 20UM -
Catalog No. 50871400
Encompass_Preferred
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with stainless-steel mesh; 3 in. dia.; No. 635; Sieve opening; 20um
Catalog No. 50-871-400 Supplier Gilson Company Inc Supplier No. GAA52
Add to Cart
Add to Cart
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with stainless-steel mesh; 3 in. dia.; No. 635; Sieve opening; 20um