Learn More
Gilson Company Inc SIEVE ACRYLIC 3 SS 32UM -
Catalog No. 50871398
Encompass_Preferred
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with stainless-steel mesh; 3 in. dia.; No. 450; Sieve opening; 32um
Catalog No. 50-871-398 Supplier Gilson Company Inc Supplier No. GAA50
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with stainless-steel mesh; 3 in. dia.; No. 450; Sieve opening; 32um