Learn More
Gilson Company Inc SIEVE ACRYLIC 3 SS NO12 -
Catalog No. 50871375
Encompass_Preferred
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with stainless-steel mesh; 3 in. dia.; No. 12; Sieve opening; 1.7mm
Catalog No. 50-871-375 Supplier Gilson Company Inc Supplier No. GAA27
Add to Cart
Add to Cart
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with stainless-steel mesh; 3 in. dia.; No. 12; Sieve opening; 1.7mm