Learn More
Gilson Company Inc SIEVE ACRYLIC 3 SS NO120 -
Catalog No. 50871390
Encompass_Preferred
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with stainless-steel mesh; 3 in. dia.; No. 120; Sieve opening; 125um
Catalog No. 50-871-390 Supplier Gilson Company Inc Supplier No. GAA42
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with stainless-steel mesh; 3 in. dia.; No. 120; Sieve opening; 125um