Learn More
Gilson Company Inc SIEVE ACRYLIC 3 SS NO16 -
Catalog No. 50871377
Encompass_Preferred
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with stainless-steel mesh; 3 in. dia.; No. 16; Sieve opening; 1.18mm
Catalog No. 50-871-377 Supplier Gilson Company Inc Supplier No. GAA29
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with stainless-steel mesh; 3 in. dia.; No. 16; Sieve opening; 1.18mm