Learn More
Gilson Company Inc SIEVE ACRYLIC 3 SS NO200 -
Catalog No. 50871393
Encompass_Preferred
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with stainless-steel mesh; 3 in. dia.; No. 200; Sieve opening; 75um
Catalog No. 50-871-393 Supplier Gilson Company Inc Supplier No. GAA45
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with stainless-steel mesh; 3 in. dia.; No. 200; Sieve opening; 75um