Learn More
Gilson Company Inc SIEVE ACRYLIC 3 SS NO230 -
Catalog No. 50871394
Encompass_Preferred
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with stainless-steel mesh; 3 in. dia.; No. 230; Sieve opening; 63um
Catalog No. 50-871-394 Supplier Gilson Company Inc Supplier No. GAA46
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with stainless-steel mesh; 3 in. dia.; No. 230; Sieve opening; 63um