Learn More
Gilson Company Inc SIEVE ACRYLIC 3 SS NO3 1/2 -
Catalog No. 50871368
Encompass_Preferred
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with stainless-steel mesh; 3 in. dia.; No. 3-1/2; Sieve opening; 5.6mm
Catalog No. 50-871-368 Supplier Gilson Company Inc Supplier No. GAA20
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with stainless-steel mesh; 3 in. dia.; No. 3-1/2; Sieve opening; 5.6mm