Learn More
Gilson Company Inc 3in (76mm) diameter Acrylic Frame Test Sieve, No. 30
Catalog No. 50871381
Encompass_Preferred
3in (76mm) diameter Acrylic Frame Test Sieve has ASTM E11 stainless steel woven-wire cloth with No. 30 (600µm) openings. Features: Required for the GA-6 GilSonic 3in AutoSiever and recommended as an alternative to metal-frame sieves for the SS-3 Gilson Performer III 3in Sieve Shaker. Clear frames make it easy to observe sample during testing, Short stacking height enables twice as many sieves in a stack compared to metal frame sieves, Compliant with ASTM E11 requirements for Compliance Test Sieves. Included Items: 3in Acrylic Frame Test Sieve with ASTM E11 stainless steel mesh, Certificate of Conformance to ASTM E11 requirements.
Catalog No. 50-871-381 Supplier Gilson Company Inc Supplier No. GAA33
3in (76mm) diameter Acrylic Frame Test Sieve has ASTM E11 stainless steel woven-wire cloth with No. 30 (600µm) openings. Features: Required for the GA-6 GilSonic 3in AutoSiever and recommended as an alternative to metal-frame sieves for the SS-3 Gilson Performer III 3in Sieve Shaker. Clear frames make it easy to observe sample during testing, Short stacking height enables twice as many sieves in a stack compared to metal frame sieves, Compliant with ASTM E11 requirements for Compliance Test Sieves. Included Items: 3in Acrylic Frame Test Sieve with ASTM E11 stainless steel mesh, Certificate of Conformance to ASTM E11 requirements.