Learn More
Gilson Company Inc SIEVE ACRYLIC 3 SS NO35 -
Catalog No. 50871382
Encompass_Preferred
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with stainless-steel mesh; 3 in. dia.; No. 34; Sieve opening; 500um
Catalog No. 50-871-382 Supplier Gilson Company Inc Supplier No. GAA34
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with stainless-steel mesh; 3 in. dia.; No. 34; Sieve opening; 500um