Learn More
Gilson Company Inc SIEVE ACRYLIC 3 SS NO400 -
Catalog No. 50871397
Encompass_Preferred
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with stainless-steel mesh; 3 in. dia.; No. 400; Sieve opening; 38um
Catalog No. 50-871-397 Supplier Gilson Company Inc Supplier No. GAA49
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with stainless-steel mesh; 3 in. dia.; No. 400; Sieve opening; 38um