Learn More
Gilson Company Inc SIEVE ACRYLIC 3 SS NO5 -
Catalog No. 50871370
Encompass_Preferred
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with stainless-steel mesh; 3 in. dia.; No. 5; Sieve opening; 4mm
Catalog No. 50-871-370 Supplier Gilson Company Inc Supplier No. GAA22
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with stainless-steel mesh; 3 in. dia.; No. 5; Sieve opening; 4mm