Learn More
Gilson Company Inc SIEVE ACRYLIC 3 SS NO60 -
Catalog No. 50871386
Encompass_Preferred
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with stainless-steel mesh; 3 in. dia.; No. 60; Sieve opening; 250um
Catalog No. 50-871-386 Supplier Gilson Company Inc Supplier No. GAA38
Add to Cart
Add to Cart
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with stainless-steel mesh; 3 in. dia.; No. 60; Sieve opening; 250um