Learn More
Gilson Company Inc SIEVE ACRYLIC 3 SS NO8 -
Catalog No. 50871373
Encompass_Preferred
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with stainless-steel mesh; 3 in. dia.; No. 8; Sieve opening; 2.36mm
Catalog No. 50-871-373 Supplier Gilson Company Inc Supplier No. GAA25
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with stainless-steel mesh; 3 in. dia.; No. 8; Sieve opening; 2.36mm