Learn More
Gilson Company Inc SIEVE ACRYLIC 3 SS NO80 -
Catalog No. 50871388
Encompass_Preferred
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with stainless-steel mesh; 3 in. dia.; No. 80; Sieve opening; 180um
Catalog No. 50-871-388 Supplier Gilson Company Inc Supplier No. GAA40
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with stainless-steel mesh; 3 in. dia.; No. 80; Sieve opening; 180um