Learn More
Marlin Steel Wire Products Electropolished Sieve Basket 18"L x 11.25"W x 4.5"H
SDP
Catalog No. NC1962281
Encompass_Preferred
Rectangular sieve basket with handles for easy gripping.Robust quality that is ideal for washing parts, degreasing, ultrasonic cleaning, curing, and material handling in a variety of applications such as chemical bath, heat treating, cryogenics, and food processing.Nestable Basket12 mesh openings per linear inch
Catalog No. NC1962281 Supplier Marlin Steel Wire Products Supplier No. 0204400131
Rectangular sieve basket with handles for easy gripping.Robust quality that is ideal for washing parts, degreasing, ultrasonic cleaning, curing, and material handling in a variety of applications such as chemical bath, heat treating, cryogenics, and food processing.Nestable Basket12 mesh openings per linear inch