Learn More
Cole-Parmer Endecotts ZSICSA-.090 Sieve Calibration Standard, soda-lime glass, 170 mesh
Catalog No. NC1639629 Shop All Cole-Parmer Products
Encompass
Soda-lime glass microspheres pass over the entire surface of the sieve and through every aperture. Standards include a calibration chart.
Catalog No. NC1639629 Supplier Cole-Parmer Supplier No. 5994519
Soda-lime glass microspheres pass over the entire surface of the sieve and through every aperture. Standards include a calibration chart.