Learn More
W.S. TYLER™ Sieve Certification and Recertification Services
Catalog No. 0490916 Shop All W. S. Tyler Products
Change view
Click to view available options
Product Type:
Certification Service
Catalog No. Product Type
04-909-16 Certification Service
1 options
Catalog No. 04-909-16 Supplier W.S. TYLER™ Supplier No. 9998
Add to Cart
Add to Cart
Certification services offered for new or used sieves
Certification Service
  • Order when purchasing new sieves
  • Uses a computerized video imaging system to verify that each sieve opening meets ASTM™ and ISO specifications within a tolerance of ±0.1 microns
  • Sieves are then serialized and supplied with documentation, traceable to NIST™, detailing the openings and the wire diameters of the mesh
Recertification Services
  • Offer the same examination and documentation processes for in-use sieves as for new sieves
  • Contact your Fisher Scientific Customer Service Representative for the recommended sieve aperture/recertification schedule
Specifications
For Use With (Equipment) New sieves
Product Type Certification Service
For Use With (Application) To serialize and supply with documentation