Learn More
Preiser Scientific Sieve Cleaning Brush, Fine Sieves, Nylon Bristle
SDP
Catalog No. NC9473346
Encompass_Preferred
Brush, Sieve Cleaning, Nylon Bristle, A nylon bristle brush recommended for fine mesh testing sieves, finer than 100 mesh, The nylon bristles will not scratch or distort the opening of the fragile fine mesh sieves
Catalog No. NC9473346 Supplier Preiser Scientific Supplier No. 90320001
Brush, Sieve Cleaning, Nylon Bristle, A nylon bristle brush recommended for fine mesh testing sieves, finer than 100 mesh, The nylon bristles will not scratch or distort the opening of the fragile fine mesh sieves