Learn More
Hogentogler & Co Inc Nylon sieve cleaning brush for 120 mesh or finer
SDP
Catalog No. NC1393236
Encompass
The 8577 nylon sieve brush is used for cleaning sieves with a 120 mesh or finer. The brush is constructed with a wood handle and a round brush head with nylon bristles.
Catalog No. NC1393236 Supplier Hogentogler & Co Inc Supplier No. 8577
The 8577 nylon sieve brush is used for cleaning sieves with a 120 mesh or finer. The brush is constructed with a wood handle and a round brush head with nylon bristles.