Learn More
Elma™ Sieve Holder for Elmasonic S50R Ultrasonic Sieve Cleaner

Description
- For holding laboratory sieves
- Hold sieves of diameter from 100 to 200mm
Specifications
Specifications
| Product Type | Sieve Holder |
| For Use With (Equipment) | Ultrasonic Cleaner, Elmasonic S50R |
| For Use With (Application) | Holds Laboratory Sieves with a Diameter of 100 to 200 mm |
| Material | Stainless Steel |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.